Learn More
ATCC Pseudomonas aeruginosa
Supplier: ATCC 19429
Pseudomonas aeruginosa strain NCTC 6750 is a whole-genome sequenced bacterial strain that was isolated from urine. This strain produces pyocyanin, pyorubin, and pyoverdin, and it has applications in testing antimicrobial preservatives and opportunistic pathogen research.