Learn More
ATCC Neisseria sicca
Catalog No. 502380146
Encompass
Neisseria sicca strain 6b is a whole-genome sequenced bacterial type strain that was isolated from the pharyngeal mucosa of a healthy man. This strain is cited to produce restriction endonuclease NsiI.
Catalog No. 50-238-0146 Supplier ATCC Supplier No. 29256
Neisseria sicca strain 6b is a whole-genome sequenced bacterial type strain that was isolated from the pharyngeal mucosa of a healthy man. This strain is cited to produce restriction endonuclease NsiI.