Learn More
ATCC Salmonella enterica; subsp. enterica; serovar Saintpaul
Catalog No. 502380151
Encompass
Salmonella enterica subsp. enterica serovar Saintpaul strain 127 was isolated from a case of cystitis in Panama. This strain is serotype 1,4,5,12:e,h:1,2 and has applications in enteric disease research.
Catalog No. 50-238-0151 Supplier ATCC Supplier No. 9712
Salmonella enterica subsp. enterica serovar Saintpaul strain 127 was isolated from a case of cystitis in Panama. This strain is serotype 1,4,5,12:e,h:1,2 and has applications in enteric disease research.