Learn More
ATCC Pseudomonas aeruginosa
Supplier: ATCC 10145
This strain of Pseudomonas aeruginosa is a whole-genome sequenced bacterial type strain. This product can be used as a positive control for molecular detection in bioaerosols, as a quality control strain for Sensititre products, or in bacterial resistance testing of adhesives and paints.