Learn More
ATCC Pseudomonas aeruginosa
Supplier:  ATCC 15692
Encompass
Pseudomonas aeruginosa strain 1C is a whole genome-sequenced strain that was isolated from an infected wound. It produces pyochelin and pyocyanin, and it can be used as a bacteriophage host or for opportunistic pathogen for research.
Catalog No. 50-238-2241