Learn More
Microbiologics™ Pseudomonas aeruginosa derived from ATCC™
SDP
Catalog No. 23047115 Shop All Microbiologics Products
Change view
Click to view available options
Format:
KWIK-STIK 2 Pack
Catalog No. Format
23-047-115 KWIK-STIK 2 Pack
1 options
Catalog No. 23-047-115 Supplier Microbiologics™ Supplier No. 01290P
Add to Cart
Add to Cart
BAA-3144™
Specifications
Biosafety Level 2
Form 2 self-contained units of a single organism
Format KWIK-STIK 2 Pack
Product Type Pseudomonas aeruginosa derived from ATCC