Learn More
ATCC Pseudomonas aeruginosa; genomic DNA
Supplier:  ATCC 15442D5
Encompass
Genomic DNA isolated from Pseudomonas aeruginosa strain PRD-10 (ATCC 15442). This whole-genome sequenced preparation can be used in PCR and other molecular biology applications.
Catalog No. 50-238-2944