Learn More
ATCC Pseudomonas aeruginosa; GFP
Supplier:  ATCC 10145GFP
Encompass
This clone was derived from ATCC 10145 and contains a multicopy vector encoding the green fluorescent protein (Aequorea victoria) GFPmut3. This gene is expressed under the control of the Plac promoter (Escherichia coli). This construct has been designed for Gram-negative bacteria fluorescence labeling. Ampicillin resistance gene (bla) encoded on a plasmid. Confers resistance to 300 μg/mL ampicillin.
Catalog No. 50-238-3178
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.