Learn More
ATCC Pseudomonas aeruginosa; PA01-LAC
Supplier:  ATCC 47085
Encompass
Pseudomonas aeruginosa strain PAO1-LAC is a whole-genome sequenced bacterium that can be used as a transformation host for the expression vector pUCP19 (ATCC 87110).
Catalog No. 50-238-2646