Learn More
ATCC Akkermansia muciniphila; Strain: m1-15
Catalog No. 502380936
Encompass
Akkermansia muciniphila strain m1-15 is a bacterial strain that was isolated from the cecal contents of a thirteen-lined ground squirrel (Ictidomys tridecemlineatus). This strain has mucin-dependent growth.
Catalog No. 50-238-0936 Supplier ATCC Supplier No. BAA2869
Akkermansia muciniphila strain m1-15 is a bacterial strain that was isolated from the cecal contents of a thirteen-lined ground squirrel (Ictidomys tridecemlineatus). This strain has mucin-dependent growth.