Learn More
Biotium Pseudomonas aeruginosa serotype 6C(1200/472), CF594 conjugate, 0.1mg/mL
Supplier:  Biotium BNC940240500
Encompass_Preferred
This antibody is specific for serotype 6C and does not react with other species. Pseudomonas aeruginosa is Gram-negative, aerobic, rod-shaped bacteria with unipolar motility. An opportunistic human pathogen, P. aeruginosa is also an opportunistic pathogen of plants. P. aeruginosa bacteria are clinically important because they are resistant to most antibiotics and they are capable of surviving in conditions that few other organisms can tolerate. Pseudomonas is often encountered in hospital and clinical work because it is a major cause of hospital acquired (nosocomial) infections. Its main targets are immuno-compromised individuals, burn victims, and individuals on respirators or with indwelling catheters. Additionally, these pathogens colonize the lungs of cystic fibrosis patients. P. aeruginosa is often identified by its pearlescent appearance and grape-like odor in vitro. Definitive clinical identification of P. aeruginosa includes identifying the production of both pyocyani
Catalog No. 50-252-3305