Learn More
ATCC Pseudomonas aeruginosa; Strain PGO2338
Supplier:  ATCC BAA2110
Encompass
Pseudomonas aeruginosa strain PGO2338 is a whole-genome sequenced, multidrug-resistant strain isolated from a human sputum sample. It was collected from a cystic fibrosis patient during a Phase III clinical trial that tested the effectiveness of aerosolized Tobi (Tobramycin). It has applications in antimicrobial resistance research and drug development.
Catalog No. 50-238-2526
 Restricted Item