Learn More
ATCC Pseudomonas aeruginosa; Strain HER-1018
Catalog No. 502384497
Encompass
Pseudomonas aeruginosa strain HER-1018 [PAO1] is a whole-genome sequenced bacterium. The strain is a permissive host for bacteriophage ATCC BAA-47-B1.
Catalog No. 50-238-4497 Supplier ATCC Supplier No. BAA47
Pseudomonas aeruginosa strain HER-1018 [PAO1] is a whole-genome sequenced bacterium. The strain is a permissive host for bacteriophage ATCC BAA-47-B1.