Learn More
ATCC Pseudomonas aeruginosa; Strain Boston 41501
Supplier:  ATCC 27853
Encompass
Pseudomonas aeruginosa strain Boston 41501 was isolated from blood culture. This whole-genome sequenced bacterial strain has applications in media and susceptibility testing, evaluation of Mueller-Hinton Agar, and as a quality control strain for Abbott, API, Autobac, BBL, bioMerieux Vitek, Micro-Media, MicroScan™, Roche Diagnostics, and Sensititre products.
Catalog No. 50-238-4830
 Restricted Item