Learn More
ATCC Pseudomonas aeruginosa; Strain PGO2332
Catalog No. 502381816
Encompass
Pseudomonas aeruginosa strain PGO2332 was collected from a sputum sample of a cystic fibrosis patient during a Phase III clinical trial that tested the effectiveness of aerosolized Tobi (Tobramycin). This strain is propagated aerobically in trypticase soy medium.
Catalog No. 50-238-1816 Supplier ATCC Supplier No. BAA2109
Pseudomonas aeruginosa strain PGO2332 was collected from a sputum sample of a cystic fibrosis patient during a Phase III clinical trial that tested the effectiveness of aerosolized Tobi (Tobramycin). This strain is propagated aerobically in trypticase soy medium.