Learn More
ATCC Pseudomonas aeruginosa; Strain PGO2401
Catalog No. 502383617
Encompass
Pseudomonas aeruginosa strain PGO2401 is a drug-resistant bacterium that was collected from a sputum sample of a cystic fibrosis patient during a Phase III clinical trial that tested the effectiveness of aerosolized Tobi (Tobramycin). This strain is whole-genome sequenced and has applications in antimicrobial resistance research and drug development.
Catalog No. 50-238-3617 Supplier ATCC Supplier No. BAA2114
Pseudomonas aeruginosa strain PGO2401 is a drug-resistant bacterium that was collected from a sputum sample of a cystic fibrosis patient during a Phase III clinical trial that tested the effectiveness of aerosolized Tobi (Tobramycin). This strain is whole-genome sequenced and has applications in antimicrobial resistance research and drug development.