Learn More
ATCC Pseudomonas aeruginosa; Strain PGO2364
Catalog No. 502383591
Encompass
Pseudomonas aeruginosa strain PGO2364 is a bacterial strain that was isolated from the sputum sample of a cystic fibrosis patient during a Phase III clinical trial that tested the effectiveness of aerosolized Tobi (Tobramycin). The presence of a phage has been observed in this strain.
Catalog No. 50-238-3591 Supplier ATCC Supplier No. BAA2112
Pseudomonas aeruginosa strain PGO2364 is a bacterial strain that was isolated from the sputum sample of a cystic fibrosis patient during a Phase III clinical trial that tested the effectiveness of aerosolized Tobi (Tobramycin). The presence of a phage has been observed in this strain.