Learn More
ATCC Pseudomonas aeruginosa; Strain PGO2354
Catalog No. 502383583
Encompass
Pseudomonas aeruginosa strain PGO2354 was collected from a sputum sample of a cystic fibrosis patient during a Phase III clinical trial that tested the effectiveness of aerosolized Tobi (Tobramycin). This whole-genome sequenced strain is propagated aerobically in trypticase soy medium and is used for drug development research.
Catalog No. 50-238-3583 Supplier ATCC Supplier No. BAA2111
Pseudomonas aeruginosa strain PGO2354 was collected from a sputum sample of a cystic fibrosis patient during a Phase III clinical trial that tested the effectiveness of aerosolized Tobi (Tobramycin). This whole-genome sequenced strain is propagated aerobically in trypticase soy medium and is used for drug development research.