Learn More
NEOGEN Pseudomonas Agar Base, 5 kg
SDP
Catalog No. 502015397
Encompass_Preferred
Pseudomonas Agar Base is used for the selective and differential isolation of Pseudomonas sp. using CN or CFC supplementation and is not intended for use in the diagnosis of disease or other conditions in humans. Pseudomonas Agar Base was developed to be supplemented with CN X107 (Cetrimide & Nalidixic Acid) as per ISO 16266-2:2018, or with CFC X108 (Cetrimide, Fusidate, & Cephaloridine) as per ISO 13720:2010. This base formula is a modification of King’s A Medium. The CN Supplement is recommended for the isolation of Pseudomonas aeruginosa, where the addition of Nalidixic Acid and the reduction of Cetrimide improved recovery.For more information visit https://www.neogen.com/solutions/Microbiology/pseudomonas-agar-base
Catalog No. 50-201-5397 Supplier NEOGEN Supplier No. 700004475
Pseudomonas Agar Base is used for the selective and differential isolation of Pseudomonas sp. using CN or CFC supplementation and is not intended for use in the diagnosis of disease or other conditions in humans. Pseudomonas Agar Base was developed to be supplemented with CN X107 (Cetrimide & Nalidixic Acid) as per ISO 16266-2:2018, or with CFC X108 (Cetrimide, Fusidate, & Cephaloridine) as per ISO 13720:2010. This base formula is a modification of King’s A Medium. The CN Supplement is recommended for the isolation of Pseudomonas aeruginosa, where the addition of Nalidixic Acid and the reduction of Cetrimide improved recovery.For more information visit https://www.neogen.com/solutions/Microbiology/pseudomonas-agar-base