Learn More
NEOGEN Pseudomonas Isolation Agar, 500 g
SDP
Catalog No. 502015585
Encompass_Preferred
Pseudomonas Isolation Agar is used for the isolation of Pseudomonas aeruginosa and other Pseudomonas spp. in a laboratory setting. Pseudomonas Isolation Agar is not intended for use in the diagnosis of disease or other conditions in humans. Pseudomonas Isolation Agar is based on Medium A described by King, Ward, and Raney. This medium is very useful for isolating Pseudomonas spp. Pseudomonas Isolation Agar includes Irgasan®, a potent broad spectrum antimicrobial not active against Pseudomonas spp. This medium is selective and formulated to enhanced formation of blue or blue-green pyocyanin pigment by Pseudomonas aeruginosa. The pigment diffuses into the medium surrounding growth.For more information visit https://www.neogen.com/solutions/Microbiology/pseudomonas-isolation-agar
Catalog No. 50-201-5585 Supplier NEOGEN Supplier No. 700003404
Pseudomonas Isolation Agar is used for the isolation of Pseudomonas aeruginosa and other Pseudomonas spp. in a laboratory setting. Pseudomonas Isolation Agar is not intended for use in the diagnosis of disease or other conditions in humans. Pseudomonas Isolation Agar is based on Medium A described by King, Ward, and Raney. This medium is very useful for isolating Pseudomonas spp. Pseudomonas Isolation Agar includes Irgasan®, a potent broad spectrum antimicrobial not active against Pseudomonas spp. This medium is selective and formulated to enhanced formation of blue or blue-green pyocyanin pigment by Pseudomonas aeruginosa. The pigment diffuses into the medium surrounding growth.For more information visit https://www.neogen.com/solutions/Microbiology/pseudomonas-isolation-agar