Learn More
Bioworld Pseudomonas Isolation Agar, 500 g
Catalog No. 50254356
Encompass_Preferred
Pseudomonas Isolation AgarUsed for isolating Pseudomonas and differentiating Pseudomonas aeruginosa from other pseudomonads based on pigment formationpH: 7.0Working Concentration: 45 g/LDehydrated Appearance: very light beige, homogeneous powderPrepared Appearance: light amber agarSupplement media with 20ml glycerol per liter of mediaComposition (g/L):Pancreatic Digest of Gelatin: 20g Magnesium Chloride: 1.4gPotassium Sulfate: 10gIrgasan (R): 0.025gAgar: 13.6g
Catalog No. 50-254-356 Supplier Bioworld Supplier No. 306200671
Pseudomonas Isolation AgarUsed for isolating Pseudomonas and differentiating Pseudomonas aeruginosa from other pseudomonads based on pigment formationpH: 7.0Working Concentration: 45 g/LDehydrated Appearance: very light beige, homogeneous powderPrepared Appearance: light amber agarSupplement media with 20ml glycerol per liter of mediaComposition (g/L):Pancreatic Digest of Gelatin: 20g Magnesium Chloride: 1.4gPotassium Sulfate: 10gIrgasan (R): 0.025gAgar: 13.6g