Learn More
ATCC Citrobacter koseri
Supplier:  ATCC 25408
Encompass
Citrobacter koseri strain 1791 [NCTC 10810] is a bacterial strain isolated from a throat. This strain is propagated aerobically on nutrient agar and broth.
Catalog No. 50-238-0499