Learn More
MOLTOX Molecular Toxicology Inc Pseudomonas P/F Agar, Bi-Plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948690
Encompass_Preferred
Pseudomonas P, or Tech Agar, enhances pyocyanin production by Pseudomonas and therefore used for the differentiation of pseudomonads. Pseudomonas F (Flo) Agar enhances fluorescin production by Pseudomonas and therefore used for the differentiation of pseudomonads. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-690 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S79
Pseudomonas P, or Tech Agar, enhances pyocyanin production by Pseudomonas and therefore used for the differentiation of pseudomonads. Pseudomonas F (Flo) Agar enhances fluorescin production by Pseudomonas and therefore used for the differentiation of pseudomonads. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.