Learn More
MOLTOX Molecular Toxicology Inc Pseudomonas P/Pseudomonas F/Cetrimide Agar, Tri Plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948702
Encompass_Preferred
Pseudomonas P, or Tech Agar, enhances pyocyanin production by Pseudomonas and therefore used for the differentiation of pseudomonads. Pseudomonas F (Flo) Agar enhances fluorescin production by Pseudomonas and therefore used for the differentiation of pseudomonads. Also known as Pseudosel Agar, Cetrimide Agar is used as a selective isolation and identification media for Pseudomonas aeruginosa. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-702 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S94
Pseudomonas P, or Tech Agar, enhances pyocyanin production by Pseudomonas and therefore used for the differentiation of pseudomonads. Pseudomonas F (Flo) Agar enhances fluorescin production by Pseudomonas and therefore used for the differentiation of pseudomonads. Also known as Pseudosel Agar, Cetrimide Agar is used as a selective isolation and identification media for Pseudomonas aeruginosa. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.