Learn More
ATCC Pseudomonas paraeruginosa strain R. Hugh 813 (ATCC 9027™)
Catalog No. 502383103
Encompass
Pseudomonas paraeruginosa strain R. Hugh 813 (ATCC 9027™) is a whole-genome sequenced strain isolated from an outer ear infection. It is used in media testing, water testing, and pharmaceutical QC. This strain is recommended as a test organism in USP <51>, <61>, <62>, and <71>.
Recommended companion products:
MicroQuant™ Pseudomonas paraeruginosa, high CFU (Fisher Cat. No. NC3456054, ATCC 9027-HQ-PACK™)
MicroQuant™ Pseudomonas paraeruginosa, low CFU (Fisher Cat. No. NC3456065, ATCC 9027-LQ-PACK™)
Genomic DNA (Fisher Cat No. 50-238-2879, ATCC 9027DQ™).
Catalog No. 50-238-3103 Supplier ATCC Supplier No. 9027
Pseudomonas paraeruginosa strain R. Hugh 813 (ATCC 9027™) is a whole-genome sequenced strain isolated from an outer ear infection. It is used in media testing, water testing, and pharmaceutical QC. This strain is recommended as a test organism in USP <51>, <61>, <62>, and <71>.
Recommended companion products:
MicroQuant™ Pseudomonas paraeruginosa, high CFU (Fisher Cat. No. NC3456054, ATCC 9027-HQ-PACK™)
MicroQuant™ Pseudomonas paraeruginosa, low CFU (Fisher Cat. No. NC3456065, ATCC 9027-LQ-PACK™)
Genomic DNA (Fisher Cat No. 50-238-2879, ATCC 9027DQ™).