Learn More
ATCC Pseudomonas paraeruginosa; 6 x 0.3mL
Supplier:  ATCC 9027MINIPACK
Encompass
Pseudomonas paraeruginosa strain R. Hugh 813 (ATCC 9027-MINI-PACK) consists of 6 ready-to-use vials of ATCC 9027 frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-4818
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.