Learn More
ATCC Pseudomonas paraeruginosa; genomic DNA
Catalog No. 502382923
Encompass
Genomic DNA isolated from Pseudomonas paraeruginosa strain R. Hugh 813 (ATCC 9207). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-2923 Supplier ATCC Supplier No. 9027D5
 Restricted Item
Add to Cart
Add to Cart
Genomic DNA isolated from Pseudomonas paraeruginosa strain R. Hugh 813 (ATCC 9207). This whole-genome sequenced product can be used in PCR and other molecular biology applications.