Learn More
ATCC Pseudomonas paraeruginosa
Catalog No. 502383201
Encompass
Pseudomonas paraeruginosa strain R. Hugh 813 is a whole-genome sequenced bacterial type strain that was isolated from an outer ear infection. This product is prepared as a certified reference material for use in challenging assay performance, validating or comparing test methods, and establishing sensitivity, linearity, and specificity during assay validation or implementation.
Catalog No. 50-238-3201 Supplier ATCC Supplier No. CRM9027
 Restricted Item
Add to Cart
Add to Cart
Pseudomonas paraeruginosa strain R. Hugh 813 is a whole-genome sequenced bacterial type strain that was isolated from an outer ear infection. This product is prepared as a certified reference material for use in challenging assay performance, validating or comparing test methods, and establishing sensitivity, linearity, and specificity during assay validation or implementation.