Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Pseudomonas protegens (G) derived from ATCC 17386, EZ-CFU
SDP

Catalog No. 10810315 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
20 pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-810-315 20 pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-810-315 Supplier Microbiologics™ Supplier No. 0524C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EZ-CFU delivers 10-100 CFU per 0.1 ml inoculum, following a simple 1:10 dilution step, each microorganism suspension provides up to 90 tests. These handy kits allow for fast, reliable Growth Promotion Testing. Plus, EZ-CFU microorganisms are just three passages from the reference strains.

  • Biosafety Level: 1
  • 20 pellets of a microorganism delivering less than 100 CFU per 0.1 mL and 10 vials [2.0 mL ea] of hydrating fluid. 
  • Following the dilution step, EZ-CFU™ provides over 90 inocula using the same suspension.
  • Formerly Pseudomonas fluorescens; biotype G

Specifications

ATCC® ATCC 17386
Biosafety Level 1
Content And Storage 2°C to 8°C
Format EZ-CFU
Manufacturing Origin United States
Product Type EZ-CFU
For Use With (Application) Growth Promotion Testing
Packaging Type Plastic Box
Quantity 20 pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.