Learn More
ATCC Pseudomonas putida
Catalog No. 502383666
Encompass
Pseudomonas putida [A.3.12, ATCC 23467, NCIB 9494, NCTC 10936, R.Y. Stanier 90] is a whole-genome sequenced bacterial type strain that degrades aromatic acids. This culture can be used as a bacteriophage host and as prey for Peredibacter starrii (ATCC 15145).
Catalog No. 50-238-3666 Supplier ATCC Supplier No. 12633
Pseudomonas putida [A.3.12, ATCC 23467, NCIB 9494, NCTC 10936, R.Y. Stanier 90] is a whole-genome sequenced bacterial type strain that degrades aromatic acids. This culture can be used as a bacteriophage host and as prey for Peredibacter starrii (ATCC 15145).