Learn More
ATCC Pseudomonas putida
Supplier:  ATCC 49128
Encompass
Pseudomonas putida strain AmMS 106 is a whole-genome sequenced clinical isolate that can be used as a quality control strain for MicroScan products.
Catalog No. 50-238-3929