Learn More
ATCC Pseudomonas putida; KT2440
Catalog No. 502384968
Encompass
Pseudomonas putida strain KT2440 is a whole-genome sequenced strain that can be used as a transformation host. This strain is genotype rmo- mod+.
Catalog No. 50-238-4968 Supplier ATCC Supplier No. 47054
Pseudomonas putida strain KT2440 is a whole-genome sequenced strain that can be used as a transformation host. This strain is genotype rmo- mod+.