Learn More
ATCC Quantified Genomic DNA; from Escherichia coli; strain HMS174; with ATCC 16S Tag 1
Catalog No. 502381305
Encompass
Quantitative genomic DNA from Escherichia coli with ATCC 16S Tag 1 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load. The synthetic Tag 1 allows for use as a spike-in control in assay development, library preparation, quality control, and data normalization for both 16S rRNA and shotgun metagenomics approaches. This product is also a component of MSA-1014.
Catalog No. 50-238-1305 Supplier ATCC Supplier No. 47011T1DQ
Quantitative genomic DNA from Escherichia coli with ATCC 16S Tag 1 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load. The synthetic Tag 1 allows for use as a spike-in control in assay development, library preparation, quality control, and data normalization for both 16S rRNA and shotgun metagenomics approaches. This product is also a component of MSA-1014.