Learn More
Cayman Chemical PseudomonIc ACDlIthIM sal 50mg
SDP
Catalog No. 502947113
Encompass_Preferred
An antibiotic; bacteriostatic against S. aureus (MIC = 0.05 UG/ml); active against skin wound clinical isolates of MRSA (MICs = 1-4 UG/ml); inhibits MRSA and P. aeruginosa biofilm formation in vitro; inhibits bacterial cell wall isoleucyl-tRNA synthetase; topical administration reduces the number of wound CFUs in a mouse model of MRSA skin infection at 2% v/v
Catalog No. 50-294-7113 Supplier Cayman Chemical Supplier No. 3327150 MG
An antibiotic; bacteriostatic against S. aureus (MIC = 0.05 UG/ml); active against skin wound clinical isolates of MRSA (MICs = 1-4 UG/ml); inhibits MRSA and P. aeruginosa biofilm formation in vitro; inhibits bacterial cell wall isoleucyl-tRNA synthetase; topical administration reduces the number of wound CFUs in a mouse model of MRSA skin infection at 2% v/v