Learn More
Cayman Chemical Pseudomonic Acid, Purity: ≥95%, CAS: 12650-69-0, 50 mg
SDP
Catalog No. 502964124
Encompass_Preferred
An antibiotic; bacteriostatic against S. aureus (MIC = 0.05 µg/ml); active against skin wound clinical isolates of MRSA (MICs = 1-4 µg/ml); inhibits MRSA and P. aeruginosa biofilm formation in vitro; inhibits bacterial cell wall isoleucyl-tRNA synthetase; topical administration reduces the number of wound CFUs in a mouse model of MRSA skin infection at 2% v/v
Catalog No. 50-296-4124 Supplier Cayman Chemical Supplier No. 2323850 MG
An antibiotic; bacteriostatic against S. aureus (MIC = 0.05 µg/ml); active against skin wound clinical isolates of MRSA (MICs = 1-4 µg/ml); inhibits MRSA and P. aeruginosa biofilm formation in vitro; inhibits bacterial cell wall isoleucyl-tRNA synthetase; topical administration reduces the number of wound CFUs in a mouse model of MRSA skin infection at 2% v/v