Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pseudooxynicotine-d4 hydrochloride | 764661-23-6 | MFCD12031608 | 99.0% | 255.18 | C10H12D4Cl2N2O | 5 MG
SDP

Catalog No. 5000262401
Encompass_Preferred

Pseudooxynicotine-d4 hydrochloride is the deuterium-labeled (d4) analogue of pseudooxynicotine supplied as the hydrochloride salt. It is intended for use as an isotopically labeled reference or internal standard in analytical and mass spectrometry applications. The product is supplied as a powdered material in small milligram pack sizes.

  • Deuterium-labeled internal standard for MS quantitation.
  • Hydrochloride salt for improved stability and handling.
  • High purity: 99.0%.
  • Molecular weight 255.18 g/mol; chemical formula C10H12D4Cl2N2O.
  • Available in small pack sizes suitable for trace analysis.
  • Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-62401 Supplier Medchemexpress LLC Supplier No. HY132962S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pseudooxynicotine-d4 hydrochloride is the deuterium-labeled (d4) analogue of pseudooxynicotine supplied as the hydrochloride salt. It is intended for use as an isotopically labeled reference or internal standard in analytical and mass spectrometry applications. The product is supplied as a powdered material in small milligram pack sizes.

  • Deuterium-labeled internal standard for MS quantitation.
  • Hydrochloride salt for improved stability and handling.
  • High purity: 99.0%.
  • Molecular weight 255.18 g/mol; chemical formula C10H12D4Cl2N2O.
  • Available in small pack sizes suitable for trace analysis.
  • Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.