Learn More
Cayman Chemical Pseudoprotodioscin, Purity: ≥98%, CAS: 102115-79-7, 100 mg
SDP
Catalog No. 502950870
Encompass_Preferred
A steroidal saponin; cytotoxic to A375, L929, and HeLa cancer cells (IC50s = 5.73, 5.09, and 3.32 µM, respectively); increases ERα nuclear accumulation in 3T3-L1 preadipocytes cultured in differentiation medium; inhibits LPS-induced increases in NF-κB activity in reporter assays using isolated mouse peritoneal macrophages and HUVECs; reduces atherosclerotic plaque area and inhibits increases in serum total cholesterol and triglyceride levels in ovariectomized ApoE-/- mice fed a high-cholesterol diet at 1, 2.5, and 5 mg/kg
Catalog No. 50-295-0870 Supplier Cayman Chemical Supplier No. 33215100 MG
A steroidal saponin; cytotoxic to A375, L929, and HeLa cancer cells (IC50s = 5.73, 5.09, and 3.32 µM, respectively); increases ERα nuclear accumulation in 3T3-L1 preadipocytes cultured in differentiation medium; inhibits LPS-induced increases in NF-κB activity in reporter assays using isolated mouse peritoneal macrophages and HUVECs; reduces atherosclerotic plaque area and inhibits increases in serum total cholesterol and triglyceride levels in ovariectomized ApoE-/- mice fed a high-cholesterol diet at 1, 2.5, and 5 mg/kg