Learn More
Medchemexpress LLC Pseudoprotodioscin | 102115-79-7 | 1031.18 | 20 MG
SDP
Catalog No. 5000416819
Encompass_Preferred
Pseudoprotodioscin is a furostanoside that inhibits SREBP1/2 and microRNA 33a/b levels, leading to a reduction in gene expression related to the synthesis of cholesterol and triglycerides. In Hep G2 cells, it increases ABCA1 protein and mRNA levels and promotes the efflux of ApoA-1-mediated cholesterol. It also inhibits SREBP1c and SREBP2 transcription by decreasing microRNA 33a/b levels, which reciprocally increases ABCA1 levels. In THP-1 macrophages, Pseudoprotodioscin reduces HMGCR, FAS, and ACC mRNA levels and promotes low-density lipoprotein receptor by decreasing PCSK9 levels.
  • Inhibits SREBP1/2 and microRNA 33a/b levels.
  • Reduces gene expression concerning cholesterol and triglyceride synthesis.
  • Increases ABCA1 protein and mRNA levels.
  • Promotes ApoA-1-mediated cholesterol efflux.
  • Reduces HMGCR, FAS, and ACC mRNA levels.
  • Promotes low-density lipoprotein receptor by decreasing PCSK9 levels.
Catalog No. 50-004-16819 Supplier Medchemexpress LLC Supplier No. HYN068620MG
Pseudoprotodioscin is a furostanoside that inhibits SREBP1/2 and microRNA 33a/b levels, leading to a reduction in gene expression related to the synthesis of cholesterol and triglycerides. In Hep G2 cells, it increases ABCA1 protein and mRNA levels and promotes the efflux of ApoA-1-mediated cholesterol. It also inhibits SREBP1c and SREBP2 transcription by decreasing microRNA 33a/b levels, which reciprocally increases ABCA1 levels. In THP-1 macrophages, Pseudoprotodioscin reduces HMGCR, FAS, and ACC mRNA levels and promotes low-density lipoprotein receptor by decreasing PCSK9 levels.
  • Inhibits SREBP1/2 and microRNA 33a/b levels.
  • Reduces gene expression concerning cholesterol and triglyceride synthesis.
  • Increases ABCA1 protein and mRNA levels.
  • Promotes ApoA-1-mediated cholesterol efflux.
  • Reduces HMGCR, FAS, and ACC mRNA levels.
  • Promotes low-density lipoprotein receptor by decreasing PCSK9 levels.