Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pseudoprotodioscin | 102115-79-7 | 99.2% | 1031.18 | C51H82O21 | 10 MG
SDP

Catalog No. 5000296146
Encompass_Preferred

Pseudoprotodioscin is a steroidal furostanoside research compound reported to modulate lipid-regulatory pathways by inhibiting SREBP1/2 and microRNA 33a/b, leading to reduced expression of genes involved in cholesterol and triglyceride synthesis. Reported cellular effects include increased ABCA1 expression and altered HMGCR, FAS, ACC, PCSK9, and LDL receptor regulation.

  • High purity (>99% by HPLC).
  • Molecular weight 1031.18 g/mol.
  • Chemical formula C51H82O21.
  • CAS number 102115-79-7.
  • Available in milligram quantities, including 10 mg vials.
  • Intended for research use only.

Catalog No. 50-002-96146 Supplier Medchemexpress LLC Supplier No. HYN068610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pseudoprotodioscin is a steroidal furostanoside research compound reported to modulate lipid-regulatory pathways by inhibiting SREBP1/2 and microRNA 33a/b, leading to reduced expression of genes involved in cholesterol and triglyceride synthesis. Reported cellular effects include increased ABCA1 expression and altered HMGCR, FAS, ACC, PCSK9, and LDL receptor regulation.

  • High purity (>99% by HPLC).
  • Molecular weight 1031.18 g/mol.
  • Chemical formula C51H82O21.
  • CAS number 102115-79-7.
  • Available in milligram quantities, including 10 mg vials.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.