Learn More
Cayman Chemical β-Pseudouridine, Purity: ≥95%, CAS: 1445-07-4, 10 mg
SDP
Catalog No. 502993757
Encompass_Preferred
The C-5 glycoside isomer of uridine; formed when uridine in RNA undergoes site-specific isomerization by a pseudouridine synthase enzyme; found in tRNAs from bacteria, archaea, and eukaryotes; reduces the number of X-ray-induced chromosomal aberrations in human lymphocytes isolated from whole blood in a dose-dependent manner
Catalog No. 50-299-3757 Supplier Cayman Chemical Supplier No. 2338310 MG
The C-5 glycoside isomer of uridine; formed when uridine in RNA undergoes site-specific isomerization by a pseudouridine synthase enzyme; found in tRNAs from bacteria, archaea, and eukaryotes; reduces the number of X-ray-induced chromosomal aberrations in human lymphocytes isolated from whole blood in a dose-dependent manner