Learn More
Medchemexpress LLC Pseudouridine | 1445-07-4 | 99.9% | 244.20 | 10 MG
SDP
Catalog No. 5000435930
Encompass_Preferred
Pseudouridine is an isomer of uridine and the most abundant modified nucleoside found in non-coding RNA. It plays a crucial role in fine-tuning and stabilizing regional structures in rRNA and tRNA, which are essential for their functions in mRNA decoding, ribosome assembly, processing, and translation. Pseudouridine-modified tRNA fragments show potential for inhibiting aberrant protein synthesis and are promising for research related to myelodysplastic syndrome (MDS)-related leukemia.
  • Isomer of uridine and most abundant modified nucleoside in non-coding RNA.
  • Fine-tunes and stabilizes regional structures in rRNA and tRNA.
  • Essential for mRNA decoding, ribosome assembly, processing, and translation.
  • Pseudouridine-modified tRNA fragments can inhibit aberrant protein synthesis.
  • Holds promise for research on myelodysplastic syndrome (MDS)-related leukemia.
Catalog No. 50-004-35930 Supplier Medchemexpress LLC Supplier No. HY11306110MG
Pseudouridine is an isomer of uridine and the most abundant modified nucleoside found in non-coding RNA. It plays a crucial role in fine-tuning and stabilizing regional structures in rRNA and tRNA, which are essential for their functions in mRNA decoding, ribosome assembly, processing, and translation. Pseudouridine-modified tRNA fragments show potential for inhibiting aberrant protein synthesis and are promising for research related to myelodysplastic syndrome (MDS)-related leukemia.
  • Isomer of uridine and most abundant modified nucleoside in non-coding RNA.
  • Fine-tunes and stabilizes regional structures in rRNA and tRNA.
  • Essential for mRNA decoding, ribosome assembly, processing, and translation.
  • Pseudouridine-modified tRNA fragments can inhibit aberrant protein synthesis.
  • Holds promise for research on myelodysplastic syndrome (MDS)-related leukemia.