Learn More
Medchemexpress LLC Pseudouridine | 1445-07-4 | 99.92% | 244.20 | 25 MG
SDP
Catalog No. 5000439569
Encompass_Preferred
Pseudouridine is an isomer of uridine and the most abundant modified nucleoside in non-coding RNA. It fine-tunes and stabilizes regional structures in rRNA and tRNA, maintaining their functions in mRNA decoding, ribosome assembly, processing, and translation. Pseudouridine-modified tRNA fragments can inhibit aberrant protein synthesis and hold promise for research on myelodysplastic syndrome (MDS)-related leukemia.
  • Isomer of uridine
  • Most abundant modified nucleoside in non-coding RNA
  • Fine-tune and stabilize regional structures in rRNA and tRNA
  • Maintain functions in mRNA decoding, ribosome assembly, processing, and translation
  • Modified tRNA fragments can inhibit aberrant protein synthesis
  • Hold promise for research on myelodysplastic syndrome (MDS)-related leukemia
  • For research use only
Catalog No. 50-004-39569 Supplier Medchemexpress LLC Supplier No. 25MGHY113061
Pseudouridine is an isomer of uridine and the most abundant modified nucleoside in non-coding RNA. It fine-tunes and stabilizes regional structures in rRNA and tRNA, maintaining their functions in mRNA decoding, ribosome assembly, processing, and translation. Pseudouridine-modified tRNA fragments can inhibit aberrant protein synthesis and hold promise for research on myelodysplastic syndrome (MDS)-related leukemia.
  • Isomer of uridine
  • Most abundant modified nucleoside in non-coding RNA
  • Fine-tune and stabilize regional structures in rRNA and tRNA
  • Maintain functions in mRNA decoding, ribosome assembly, processing, and translation
  • Modified tRNA fragments can inhibit aberrant protein synthesis
  • Hold promise for research on myelodysplastic syndrome (MDS)-related leukemia
  • For research use only