Learn More
Medchemexpress LLC Pseudouridine | 1445-07-4 | MFCD00038458 | 99.9% | 244.20 | 5 MG
SDP
Catalog No. 5000364398
Encompass_Preferred
Pseudouridine is an isomer of uridine and the most abundant modified nucleoside in non-coding RNA. It fine-tunes and stabilizes regional structures in rRNA and tRNA, maintaining their functions in mRNA decoding, ribosome assembly, processing, and translation. Pseudouridine-modified tRNA fragments can inhibit aberrant protein synthesis and hold promise for research on myelodysplastic syndrome (MDS)-related leukemia.
  • Most abundant modified nucleoside in non-coding RNA
  • Fine-tunes and stabilizes regional structures in rRNA and tRNA
  • Maintains functions in mRNA decoding, ribosome assembly, processing, and translation
  • Inhibits aberrant protein synthesis
  • Forms additional hydrogen bonds, stabilizing RNA and improving base stacking
  • Found in all three phylogenetic domains of life
Catalog No. 50-003-64398 Supplier Medchemexpress LLC Supplier No. 5MGHY113061
Pseudouridine is an isomer of uridine and the most abundant modified nucleoside in non-coding RNA. It fine-tunes and stabilizes regional structures in rRNA and tRNA, maintaining their functions in mRNA decoding, ribosome assembly, processing, and translation. Pseudouridine-modified tRNA fragments can inhibit aberrant protein synthesis and hold promise for research on myelodysplastic syndrome (MDS)-related leukemia.
  • Most abundant modified nucleoside in non-coding RNA
  • Fine-tunes and stabilizes regional structures in rRNA and tRNA
  • Maintains functions in mRNA decoding, ribosome assembly, processing, and translation
  • Inhibits aberrant protein synthesis
  • Forms additional hydrogen bonds, stabilizing RNA and improving base stacking
  • Found in all three phylogenetic domains of life