Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pseudouridine | 1445-07-4 | MFCD00038458 | 99.98% | 244.20 g/mol | C9H12N2O6 | 5 MG
SDP

Catalog No. 5000264191
Encompass_Preferred

Pseudouridine is an analytical reference standard of the nucleoside pseudouridine, intended for research and analytical applications. It stabilizes regional structures in rRNA and tRNA and is used as a reference material for qualitative and quantitative analyses.

  • Analytical reference standard for method development and calibration.
  • High purity for accurate quantitative results.
  • Suitable for HPLC, GC, and MS analysis.
  • Commonly supplied in small milligram quantities for laboratory use.
  • Molecular formula C9H12N2O6; molecular weight 244.20 g/mol.
  • CAS number 1445-07-4.

Catalog No. 50-002-64191 Supplier Medchemexpress LLC Supplier No. HY113061R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pseudouridine is an analytical reference standard of the nucleoside pseudouridine, intended for research and analytical applications. It stabilizes regional structures in rRNA and tRNA and is used as a reference material for qualitative and quantitative analyses.

  • Analytical reference standard for method development and calibration.
  • High purity for accurate quantitative results.
  • Suitable for HPLC, GC, and MS analysis.
  • Commonly supplied in small milligram quantities for laboratory use.
  • Molecular formula C9H12N2O6; molecular weight 244.20 g/mol.
  • CAS number 1445-07-4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.