Learn More
Cayman Chemical Pseudouridine 25mg
SDP
Catalog No. 502993780
Encompass_Preferred
The C-5 glycoside isomer of uridine; formed when uridine in RNA undergoes site-specific isomerization by a pseudouridine synthase enzyme; found in tRNAs from bacteria, archaea, and eukaryotes; reduces the number of X-ray-induced chromosomal aberrations in human lymphocytes isolated from whole blood in a dose-dependent manner
Catalog No. 50-299-3780 Supplier Cayman Chemical Supplier No. 2338325 MG
Add to Cart
Add to Cart
The C-5 glycoside isomer of uridine; formed when uridine in RNA undergoes site-specific isomerization by a pseudouridine synthase enzyme; found in tRNAs from bacteria, archaea, and eukaryotes; reduces the number of X-ray-induced chromosomal aberrations in human lymphocytes isolated from whole blood in a dose-dependent manner