Learn More
Medchemexpress LLC Pseudouridine 5'-triphosphate trisodium solution (100mM) | 1175-34-4 | 98.0% | C9H12N2Na3O15P3 | 5.5MG
SDP
Catalog No. 5000356946
Encompass_Preferred
Pseudouridine 5'-triphosphate trisodium solution (100mM) is a modified nucleoside commonly utilized in the polymerase-mediated synthesis of RNA. This compound, when incorporated into mRNA, offers improved nuclease stability, immunogenicity, and translational efficiency compared to unmodified uridine-containing mRNA. The salt form ensures enhanced water solubility and stability, making it suitable for various research applications.
  • Modified nucleoside for RNA synthesis.
  • Improves nuclease stability of mRNA.
  • Reduces immunogenicity of mRNA.
  • Enhances translational properties of mRNA.
  • Enhanced water solubility.
  • Increased stability.
  • For research use only.
Catalog No. 50-003-56946 Supplier Medchemexpress LLC Supplier No. HY141567A5.5MG
Pseudouridine 5'-triphosphate trisodium solution (100mM) is a modified nucleoside commonly utilized in the polymerase-mediated synthesis of RNA. This compound, when incorporated into mRNA, offers improved nuclease stability, immunogenicity, and translational efficiency compared to unmodified uridine-containing mRNA. The salt form ensures enhanced water solubility and stability, making it suitable for various research applications.
  • Modified nucleoside for RNA synthesis.
  • Improves nuclease stability of mRNA.
  • Reduces immunogenicity of mRNA.
  • Enhances translational properties of mRNA.
  • Enhanced water solubility.
  • Increased stability.
  • For research use only.