Learn More
Sigma Aldrich Fine Chemicals Biosciences Pseudouridine 9825MG
Catalog No. 502897568
Encompass
Pseudouridine is a C-glycosyl pyrimidine that consists of uracil having a beta-D-ribofuranosyl residue attached at position 5. The C-glycosyl isomer of the nucleoside uridine. It has a role as a fundamental metabolite. Pseudouridine is found in all species and in all classes of RNA except mRNA. It is formed by enzymes called pseudouridine synthases which post-transcriptionally isomerize specific uridine residues in RNA.
Catalog No. 50-289-7568 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SMB0091225MG
Pseudouridine is a C-glycosyl pyrimidine that consists of uracil having a beta-D-ribofuranosyl residue attached at position 5. The C-glycosyl isomer of the nucleoside uridine. It has a role as a fundamental metabolite. Pseudouridine is found in all species and in all classes of RNA except mRNA. It is formed by enzymes called pseudouridine synthases which post-transcriptionally isomerize specific uridine residues in RNA.