Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Pseudouridine, MP Biomedicals™

Catalog No. MP219917883 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219917883 250 mg
MP219917810 10 mg
MP219917850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219917883 Supplier MP Biomedicals, Inc Supplier No. 0219917883
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Used in initiation of translation. β-Pseudouridine, MP Biomedicals is an isomer of the nucleoside uridine, found in all species and in many classes of RNA (except mRNA). Studies suggest that it reduces radiation-induced chromosome aberrations in huma

β-Pseudouridine is formed by enzymes called Ψ synthases, which post-transcriptionally isomerize specific uridine residues in RNA in a process termed pseudouridylation.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-07-4
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.20
MDL Number MFCD00038458
InChI Key PTJWIQPHWPFNBW-GBNDHIKLSA-N
Synonym pseudouridine, beta-pseudouridine, psi-uridine, 5-beta-d-ribofuranosyl uracil, beta-d-pseudouridine, 5-b-d-ribofuranosyl-uracil, pseudouridine c, 5-ribosyluracil, 1s-1,4-anhydro-1-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl-d-ribitol, 5-2s,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione
PubChem CID 15047
ChEBI CHEBI:17802
IUPAC Name 5-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)C1=CNC(=O)NC1=O

Specifications

Density 1.64g/cm3
Boiling Point 598.4°C (760 mmHg (literature))
Refractive Index 1.62 (Lit.)
Quantity 250 mg
Solubility Information Soluble in water (50mg/mL, clear, colorless solution).
Formula Weight 244.2g/mol
Percent Purity 99.40%
Grade Not Available
Physical Form Powder
Recommended Storage Store at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.