Learn More
Medchemexpress LLC Pseudouridine (Standard) | 1445-07-4 | 99.98% | 244.20 | 100 MG
SDP
Catalog No. 5000435931
Encompass_Preferred
Pseudouridine (Standard) is the analytical standard of Pseudouridine, intended for research and analytical applications. It is an isomer of the nucleoside uridine and the most abundant modified nucleoside in non-coding RNAs. In rRNA and tRNA, Pseudouridine helps fine-tune and stabilize regional structure, supporting their functions in mRNA decoding, ribosome assembly, processing, and translation.
  • Analytical standard for research and analytical applications.
  • Most abundant modified nucleoside in non-coding RNAs.
  • Helps fine-tune and stabilize regional structure of rRNA and tRNA.
  • Supports mRNA decoding, ribosome assembly, processing, and translation.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
Catalog No. 50-004-35931 Supplier Medchemexpress LLC Supplier No. HY113061R100MG
Pseudouridine (Standard) is the analytical standard of Pseudouridine, intended for research and analytical applications. It is an isomer of the nucleoside uridine and the most abundant modified nucleoside in non-coding RNAs. In rRNA and tRNA, Pseudouridine helps fine-tune and stabilize regional structure, supporting their functions in mRNA decoding, ribosome assembly, processing, and translation.
  • Analytical standard for research and analytical applications.
  • Most abundant modified nucleoside in non-coding RNAs.
  • Helps fine-tune and stabilize regional structure of rRNA and tRNA.
  • Supports mRNA decoding, ribosome assembly, processing, and translation.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.