Learn More
AdipoGen Pseurotin A
SDP
Catalog No. 502562158
Encompass_Preferred
Chemical. CAS 58523-30-1. Formula C22H25NO8. MW 431.4. Isolated from Aspergillus fumigatus. Antibiotic. Chitin synthase inhibitor. Cytotoxic. Shows nematicidal activity.
Catalog No. 50-256-2158 Supplier AdipoGen Supplier No. BVT0003M005
Chemical. CAS 58523-30-1. Formula C22H25NO8. MW 431.4. Isolated from Aspergillus fumigatus. Antibiotic. Chitin synthase inhibitor. Cytotoxic. Shows nematicidal activity.